Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Smooth Latex Coated Seamless Knit Cotton/Polyester Gloves

Catalog No. 19165740 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-165-740 Large
19-165-739 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-165-740 Supplier PIP™ Supplier No. 39C122/L
  • $31.75
    $15.65 / Dozen of 12

    Save $16.10

    51% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Seamless knit shell is comfortable and flexible, with a latex coated palm for abrasion resistance and enhanced grip capabilities.

  • Latex coating enhances grip in wet/dry conditions
  • Seamless cotton/polyester knit shell is comfortable and flexible
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Coating on the palm/fingers provides abrasion resistance
  • Breathable back for comfort

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Latex
Color Natural, Blue
Exterior Finish No Texture
Grade Regular
Hand Specific
Material Polycotton
Product Type Seamless Knit Gloves
Size Large
Style Standard
For Use With (Application) Assembly, Construction, Inspection, Material Handling, and Landscaping
Glove Cuff Style Continuous Knit
Series 39-C122
Length (English) 9.8 in.
Length (Metric) 25 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.