Learn More
PIP™ Eva™ Women's Pink Safety Glasses
Catalog No. 19823990 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Frame Color:
Clear, Pink
Gray, Pink
Lens Coating:
Anti-fog, Scratch-resistant
Hardcoat
Lens Tint:
Clear
Gray
Pink
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
19-823-990 Clear, Pink Hardcoat Clear
19-823-991 Clear, Pink Anti-fog, Scratch-resistant Clear
19-823-993 Gray, Pink Hardcoat Gray
19-823-992 Clear, Pink Hardcoat Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-823-990 Supplier PIP™ Supplier No. 250100900
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specially designed for women's facial structures
Feature a Pink Ribbon on frames; a portion of the proceeds from the purchase of this product will go to help breast cancer research.
  • Polycarbonate lenses block 99.9% of UV A/B radiation
  • Smaller vertical size fits on a woman's higher cheekbones
  • Rimless frame, rubber temple ends, polycarbonate bridge
  • Soft rubber temples offer a comfortable and secure fit
  • Lightweight construction ensures all-day comfort
  • Meets ANSI Z87.1+ and CSA Z94.3
  • 10 Base curve lens
Used for general manufacturing and anywhere eye protection is needed.
Compliance
Meets ANSI Z87.1-2015 standards.
Specifications
Lens Coating Hardcoat
Certifications/Compliance ANSI Z87, CSA Z94.1
Lens Tint Clear
Product Type Women's Safety Glasses
For Use With (Application) General manufacturing
Frame Color Clear, Pink
Temple Style Wraparound
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
Frame Type Rimless
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.