Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ 3GX™ 18-Gauge Dyneema™ Diamond Blended Gloves, Polyurethane Coating

Catalog No. 19153125 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Glove Cuff Color:
Black
Blue
Brown
Purple
Red
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
19-153-125 2X-Large Blue
19-153-120 X-Small Purple
19-153-121 Small Red
19-153-122 Medium Yellow
19-153-123 Large Brown
19-153-124 X-Large Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-153-125 Supplier PIP™ Supplier No. 19D318/XXL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Lightweight shell with polyurethane coating provides dexterity, tactile sensitivity and ANSI A2 cut resistance. Gloves are also washable, resistant to chemicals, water and ultraviolet light.

  • Seamless knit, Dyneema™ diamond blended
  • Polyurethane coated smooth grip
  • Washable, resistant to chemicals, water and UV light
  • Color coded cuff hems for easy identification
  • Puncture and abrasion resistance
  • Cut-resistant and tactile sensitivity
  • Applications include electronics, glass, metal, food processing

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) CE
Construction Seamless Knit
Coating Material Polyurethane
Color Black, Blue
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free Yes
Material Dyneema, Lycra, Polyurethane
Product Type Cut-resistant Gloves
Size 2X-Large
EN388 Standard EN 388:3342
For Use With (Application) For applications that require exceptional dexterity, tactile sensitivity and cut resistance
Glove Cuff Color Blue
Glove Cuff Style Knit
Hem Color Light Blue
Model 19-D318/XXL
Puncture Rating ANSI/ISEA 105-16 Level 2
Series 19-D318
Yarn Gauge 18
Length (English) 10.2 in.
Length (Metric) 26 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.