Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ Crinkle Latex Coated Seamless Knit Cotton/Polyester Gloves

Catalog No. 19825286 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-825-286 X-Large
19-825-282 X-Small
19-825-283 Small
19-825-284 Medium
19-825-285 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-825-286 Supplier PIP™ Supplier No. 39C1305/XL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Seamless knit shell is comfortable and flexible, while the latex crinkle coating is specially designed to channel fluids away from the glove for better contact on wet or dry surfaces.

  • Seamless cotton/polyester knit shell is breathable, comfortable, and flexible
  • Latex crinkle coating features creases on the surface that are designed to channel fluids away for improved contact on wet or dry surfaces
  • Coating is flexible, tough and durable due to its high degree of resistance to snagging, puncture, and abrasion
  • Knit Wrist helps prevent dirt and debris from entering the glove
  • Ergonomic shaped curved finger pattern for increased user comfort

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Crinkled Latex
Color Blue, Gray
Exterior Finish Fully Textured
Grade Regular
Hand Specific
Material Polycotton
Product Type Seamless Knit Gloves
Size X-Large
Style Standard
For Use With (Application) Assembly, Construction, Inspection, Material Handling, and Landscaping
Glove Cuff Style Continuous Knit
Series 39-C1305
Yarn Gauge 10
Length (English) 11 in.
Length (Metric) 28 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.