Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ CR Plus Polyurethane Coated Dyneema™ Fiber Gloves

Catalog No. 19151231 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
2X-Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-151-231 2X-Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-151-231 Supplier PIP™ Supplier No. 19D625/XXS
This item is not returnable. View return policy

Dyneema™ fiber blend provides a high level of protection against cuts and abrasions, while the polyurethane coating enhances grip capabilities.

  • Dyneema™, and Nylon Lycra™ shell provides dexterity, tactile sensitivity and cut resistance
  • Polyurethane coating on the palm and fingertips offers abrasion resistance, puncture resistance, and enhanced wet/dry grip
  • Knit wrist helps prevent dirt and debris from entering glove
  • Resistant to chemicals, water and ultraviolet light
  • Washable

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Polyurethane
Color White
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Material Dyneema, Lycra
Product Type Seamless Knit Gloves
Size 2X-Small
For Use With (Application) Electronics, Glass Cutting, Metal Handling, and Food Processing
Glove Cuff Style Continuous Knit
Series 19-D625
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.