Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ CR Plus Gloves

Catalog No. 19151252 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-151-252 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-151-252 Supplier PIP™ Supplier No. 19D650/L
This item is not returnable. View return policy

Medium weight, coated gloves made with Dyneema™

Polyethylene fibers 10 to 15 times stronger than steel, more durable than polyester and 40% stronger than aramid fiber. Used for electronics, glass cutting, metal handling, food processing and meat processing.

  • Gray spun Dyneema yarn knitted shell (13 gauge) with nylon/Lycra™
  • Gray polyurethane-coated palm and finger tips
  • Washable, resistant to chemicals, water and ultraviolet light
  • Seamless construction
  • Color-coded hems for easy size identification

Specifications

Color Gray
Cut Resistance Rating ANSI A3
Material Dyneema, Nylon, Lycra
Product Type Cut-resistant Gloves
Size Large
For Use With (Application) Electronics, glass cutting, metal handling, food processing and meat processing
Glove Cuff Color Brown
Glove Cuff Style Continuous Knit
Hem Color Brown
Length (English) 9.8 in.
Length (Metric) 25 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.