Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ CR Ultra Seamless Knit Dyneema™ Gloves with Crinkle Latex Coating

Catalog No. 19151278 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-151-278 Large
19-151-277 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-151-278 Supplier PIP™ Supplier No. 19D813/L
This item is not returnable. View return policy

Seamless knit construction is comfortable and breathable, with Dyneema™ Fibers for tactile sensitivity and cut resistance. Crinkle latex coating enhances grip capabilities in wet or dry conditions.

  • Seamless construction increases comfort and breathability
  • Dyneema™ shell is lightweight to provide dexterity, tactile sensitivity and cut resistance
  • Crinkle latex coating enhances grip capabilities in wet/dry conditions
  • Knit Wrist helps prevent dirt and debris from entering the glove
  • Resistant to chemicals, water, and ultraviolet light
  • Washable

Specifications

Construction Seamless Knit
Coating Material Crinkled Latex
Color Blue
Cut Resistance Rating ANSI A4
Exterior Finish Fully Textured
Hand Specific
Material Dyneema Fiber
Product Type Cut-resistant Gloves
Size Large
For Use With (Application) Electronics, glass cutting, metal handling, and food processing
Glove Cuff Color White
Glove Cuff Style Continuous Knit
Series 19-D813
Length (English) 9.8 in.
Length (Metric) 25 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.