Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ Nitrile Coated Seamless Knit Cotton/Polyester Gloves

Catalog No. 19165734C Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-165-734C Large
19-165-734B Medium
19-165-734A Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-165-734C Supplier PIP™ Supplier No. 381410/L

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Seamless cotton/polyester shell is comfortable and flexible, while the nitrile coating is highly impermeable to liquids for secure gripping power.

  • Nitrile coating is highly impermeable to liquids
  • Provides a tacky dry grip for specific applications that require secure holding power
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Color coded hems for easy size identification
  • Ergonomic shaped curved finger pattern
  • Breathable glove shell

Specifications

Coated Area Palm
Construction Seamless Knit
Coating Material Nitrile
Color Gray, Black
Exterior Finish No Texture
Hand Specific
Material Polycotton
Product Type Seamless Knit Gloves
Size Large
For Use With (Application) Automotive Plants, Material Handling, Packaging, Construction, and Warehousing
Glove Cuff Style Continuous Knit
Series 38-1410
Length (English) 10 in.
Length (Metric) 25.5 cm
Show More Show Less

WARNING: Cancer - www.P65Warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.