Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ MaxiFlex™ Ultimate™ Nitrile Coated Nylon Gloves with Partial MicroFoam Grip

Catalog No. p-2912470 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
2X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-150-1638 2X-Small
19-130-1178B Small
19-130-1178C Medium
19-130-1178D Large
19-130-1178E X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-150-1638 Supplier PIP™ Supplier No. 34875/XXS
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

MicroFoam Nitrile coating is compatible with light oils to provide abrasion resistance, and enhanced grip capabilities. Gloves are touchscreen compatible so that users can stay connected to phones and other touch screen devices.

  • Knit wrist helps prevent dirt and debris from entering the gloves
  • Patented micro-foam nitrile coating offers 360° breathability
  • 25% thinner than typical nitrile gloves while offering twice the mechanical performance
  • Glove design mimics the hand at rest to reduce hand fatigue and increase comfort
  • Constructed with the latest knitting technology to produce smooth and rounded fingertips, improving fingertip sensitivity
  • Micro-cup finish allows for a controlled grip in a variety of conditions
  • Gloves are laundered prior to final packaging to ensure that they are clean and safe for use
  • All ingredients used in the manufacturing and construction of this product are compliant with REACH legislation
  • Oeko-Tex™ Association has evaluated and certified this product as skin safe from the second it comes into contact with the skin
  • Skin Health Alliance has awarded professional dermatological accreditation following review of the scientific dossier behind this product

Specifications

ANSI Abrasion Rating ANSI 3
Construction Seamless Knit
Coating Material Nitrile Foam
Color Black
Exterior Finish No Texture
Hand Specific
Material Nylon, Spandex
Product Type Seamless Knit Gloves
Size 2X-Small
Style Standard
EN388 Standard 4131
For Use With (Application) General Assembly, Micro-Engineering, Painting, and Horticulture
Glove Cuff Color Gray
Glove Cuff Style Continuous Knit
Series 34-875
Yarn Gauge 15
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.