Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ MaxiFlex™ Ultimate™ Nitrile Coated Nylon Lycra™ Gloves with Full MicroFoam Grip

Catalog No. p-3080881 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-160-834C Small
19-160-834D Medium
19-160-834E Large
19-160-834F X-Large
19-160-834G 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-160-834C Supplier PIP™ Supplier No. 34876/S
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Micro-Foam Nitrile coating is compatible with light oils to provide abrasion resistance, and enhanced grip capabilities. Designed and developed as a breathable glove, the MaxiFlex™ Ultimate™ is ideal for precision handling in dry environments.

  • Knit wrist helps prevent dirt and debris from entering the glove
  • Patented micro-foam nitrile coating offers 360° breathability
  • 25% thinner than most foam nitrile gloves on the market, while also offering twice the mechanical performance
  • Glove design mimics the hand at rest to reduce hand fatigue and increase comfort
  • Constructed with the latest knitting technology to produce smooth and rounded fingertips
  • Consistent liner penetration from the nitrile, leaving only the soft liner resting against the skin
  • Micro-cup finish allows for a controlled grip
  • Gloves are laundered prior to final packaging to ensure they are clean and safe for use
  • Complies with FDA food handling requirements 21 CFR, Part 177
  • All ingredients used in the manufacturing and construction of this product are compliant with the REACH legislation
  • Oeko-Tex™ Association has evaluated and certified this product as skin safe from the second it comes into contact with the skin
  • Skin Health Alliance has awarded professional dermatological accreditation following review of the scientific dossier behind this product
  • 100% silicone free

Specifications

Coated Area Full Glove
Coating Material Nitrile MicroFoam
Color Black
Exterior Finish Fully Textured
Hand Specific
Material Nylon, Spandex
Product Type Seamless Knit Gloves
Size Small
Style Standard
For Use With (Application) General Assembly, Micro-Engineering, Automotive Components, Horticulture, and Painting
Glove Cuff Color Gray
Glove Cuff Style Continuous Knit
Series 34-876
Yarn Gauge 15
Length (English) 8.3 in.
Length (Metric) 21 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.