Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ MaxiFoam™ Premium Seamless Knit Nylon Gloves with Nitrile Foam Coating

Catalog No. 191301174D Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-1174D Large
19-130-1174C Medium
19-130-1174E X-Large
19-130-1174B Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-130-1174D Supplier PIP™ Supplier No. 34800/L
  • $2,191.00
    $1,091.65 / Case of 144

    Save $1,099.35

    50% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nitrile MicroFoam coating is compatible with light oils to provide abrasion resistance and enhance grip capabilities. Seamless knit nylon construction increases comfort, finger dexterity, and breathability.

  • Surface of the glove is designed to channel oil away for an improved grip
  • Minimal strike-through reduces irritation and increases comfort levels
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Designed to produce minimal lint and dust, 100% silicone free
  • Gloves are laundered prior to final packaging to ensure that they are clean and safe for use
  • Oeko-Tex™ Association has evaluated and certified this product as skin safe
  • Complies with FDA Food Handling Requirements 21 CFR, Part 177

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile Foam
Color White
Exterior Finish Fully Textured
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Large
Style Standard
For Use With (Application) General Assembly, Micro-Engineering, Painting, and Horticulture
Glove Cuff Color Natural
Glove Cuff Style Continuous Knit
Series 34-800
Yarn Gauge 15
Length (English) 8.7 in.
Length (Metric) 22 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.