Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ GP™ White Polyurethane Coated Seamless Knit Nylon Gloves

Catalog No. 19149574 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-149-574 X-Large
19-149-570 X-Small
19-149-573 Large
18-150-0851 2X-Large
19-149-571 Small
19-149-572 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-149-574 Supplier PIP™ Supplier No. 33125/XL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Seamless knit nylon shell increases comfort, finger dexterity, and breathability. Polyurethane coating provides a tactile grip in dry and slightly oily conditions.

  • Seamless knit nylon shell increases comfort, finger dexterity, and breathability
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Breathable, nylon back increases user comfort
  • Ideal for applications that require dexterity for precise handling
  • Packed in pairs for ease of distribution

Specifications

ANSI Abrasion Rating ANSI 1
Construction Seamless Knit
Coating Material Polyurethane
Color White
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size X-Large
Style Standard
EN388 Standard 4131AX
For Use With (Application) Electronics/Computer Assembly, Quality Control, Inspection and General Assembly
Glove Cuff Style Continuous Knit
Series 33-125
Length (English) 9.6 in.
Length (Metric) 24.5 cm
Show More Show Less

WARNING: Cancer - www.P65Warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.