Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Premium Grade Cotton Chore Gloves with Double Layer Palm/Back

Catalog No. 191201607 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-120-1607 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-120-1607 Supplier PIP™ Supplier No. 93589
This item is not returnable. View return policy

Cotton construction keeps hands cool and protected, while the double palm provides additional weight, durability, and protection. This multi-purpose glove can be used for light protection in warehousing, farming, and automotive applications.

  • Canvas Back offers breathability to the back of the hand
  • Band Top cuff helps protect wrist area and allows for easy on and off
  • Nap-out finish offers moderate heat resistance
  • Straight thumb increases comfort in closed-fisted work
  • Clute cut provides a roomy fit

Specifications

Back Material Canvas
Cuff Material Canvas
Color Gold, Natural
Exterior Finish Nap-out
Grade Premium
Hand Specific
Material Cotton
Product Type Chore Gloves
Size Large
Style Clute
Gender Men's
Glove Cuff Color Natural
Glove Cuff Style Band Top
Hem Color Red
Palm Type Double
Thumb Style Straight
Yarn Weight 16 oz.
Length (English) 9.5 in.
Length (Metric) 24.1 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.