Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Maximum Safety™ FR Treated Synthetic Leather Palm Utility Gloves

Catalog No. 19103611 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-103-611 3X-Large
19-103-608 Large
19-103-609 X-Large
19-103-610 2X-Large
19-103-606 Small
19-103-607 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-103-611 Supplier PIP™ Supplier No. 731703/XXXL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Aramid Fiber liner provides cut protection and heat dispersion. FR treated synthetic suede leather palm and finger tips increase heat protection.

  • Aramid Fiber lining provides comfort, enhanced cut protection, and good heat dispersion
  • FR treated synthetic suede leather palm and finger tips for heat protection
  • Fourchettes between the fingers enhance the natural contour of the glove for a better fit
  • Reinforced thumb crotch and cushioned knuckles for additional durability in high-wear areas
  • Double palm offers comfort, insulation, and heat protection
  • Elastic closure sewn inside glove for safety and comfort
  • Extended slip-on cuff allows for easy on/off and forearm protection

Specifications

Color Black
Cut Resistance Rating ANSI A3
Hand Specific
Material Suede Leather
Size 3X-Large
For Use With (Application) Hot/Heavy Material Handling, Stamping, Grinding, and Welding
Glove Cuff Style Open
Liner Material Aramid Fiber
Series 73-1703
Length (English) 11 in.
Length (English) Cuff 2.5 in.
Length (Metric) 28 cm
Length (Metric) Cuff 6.35 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.