Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Premium Grade Split Cowhide Leather Gloves with Fleece Pile Lining

Catalog No. 19013569 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
2X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-013-569 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-013-569 Supplier PIP™ Supplier No. 77289/XXL
This item is not returnable. View return policy

Split Cowhide leather construction provides comfort, durability, and abrasion resistance. Fleece pile lining is an extra thick, synthetic fleece fabric that provides superior warmth in cold applications.

  • Cowhide leather construction provides comfort, durability, abrasion resistance, and breathability
  • Fleece pile lining is an extra thick, synthetic fleece fabric that provides superior warmth
  • Drivers gloves feature a slip-on design to provide good fit and comfort
  • Keystone thumb is ergonomic, reduces hand fatigue, offers superior dexterity and durability
  • Elastic closure sewn inside glove for safety and comfort

Specifications

Color Natural
Hand Specific
Material Cowhide Leather
Product Type Driver Gloves
Size 2X-Large
For Use With (Application) Farming, Construction, Machine Operation, and Refrigeration
Glove Cuff Style Open
Liner Material Fleece Pile
Series 77-289
Thumb Style Keystone
Length (English) 11 in.
Length (Metric) 28 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.