Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Regular Grade Split Cowhide Leather Gloves with Thermal Lining

Catalog No. p-3348810 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-822-676 Large
19-822-677 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-822-676 Supplier PIP™ Supplier No. 77238/L
This item is not returnable. View return policy

Split Cowhide leather construction provides comfort, durability, and abrasion resistance. White thermal lining provides insulation.

  • Split leather is economical and has a visible texture
  • Drivers gloves feature slip-on styling to provide good fit and comfort
  • Straight thumb increases comfort in closed-fisted work
  • Elastic closure sewn inside glove for safety and comfort
  • Cotton color binding denotes sizing

Specifications

Color Tan
Grade Regular
Hand Specific
Material Shoulder Split Cowhide Leather
Product Type Thermal Lined Gloves
Size Large
For Use With (Application) Farming, Construction, Machine Operation, Refrigeration, and Cold Weather Applications
Glove Cuff Style Open
Hem Color Brown
Lining Thermal Lining
Series 77-238
Thumb Style Straight
Length (English) 10.3 in.
Length (Metric) 26 cm
Weight 30.8 lb. (Case)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.