Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Assurance™ 48-L185T Unsupported Latex Glove, Flock Lining

Catalog No. 19060035 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Orange; Large; Thickness: 18mil.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-060035 Orange; Large; Thickness: 18mil.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19-060035 Supplier PIP™ Supplier No. 48L185T/L

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Flock lined, 18 mil glove features a honeycomb pattern for an enhanced grip in low hazard areas.

Used in janitorial, food handling and processing, laboratory, manufacturing and maintenance

  • Medium weight; recessed diamond grip
  • Chemical resistant; unsupported latex
  • Length: 12 in. (30.5 cm)
  • Thickness: 18 mil
  • Color: Orange

Specifications

Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) EN 374
Color Orange
Finger Thickness (English) 18 mil
Finger Thickness (Metric) 0.46 mm
Latex Free No
Material Latex
Palm Thickness (English) 18 mil
Palm Thickness (Metric) 0.46 mm
Size Large
Thickness (English) 18 mil
Thickness (Metric) 0.46 mm
EN 421 No
EN ISO 374-1 CHEMICAL LETTER CODES AKL
For Use With (Application) Janitorial, sanitation and general maintenance
Glove Cuff Style Beaded/Rolled
Grip Design Honeycomb
Liner Material Cotton Flock
Lining Lined
Manufacturing/QMS Audit Standards ISO 9001:2000
Model 48-L185T/L
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.