Learn More
PIP™ Marigold Unsupported Latex Gloves, Unlined
Catalog No. 19100557 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Large
Catalog No. Size
19-100-557 Large
1 options
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19-100-557 Supplier PIP™ Supplier No. 472242T/L
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Premium unlined, medium-weight natural rubber latex provides excellent wear protection and flexibility
Used in electronics, general assembly, laboratory, janitorial, manufacturing and maintenance
  • Fluted cuff design enables ventilation to the hand while providing maximum protection
  • Beaded, tear-resistant cuff for ease of donning
  • Unique roughened grip pattern offers good grip in damp conditions
  • Length: 13 in. (33cm)
  • Thickness: 18mil
  • Color: Orange
Specifications
Material Latex
Size Large