Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ MaxiFlex™ Active™ Nitrile Coated Nylon Lycra™ Gloves with MicroFoam Grip

Catalog No. 19131605 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-131-605 X-Large
19-131-600 2X-Small
19-131-601 X-Small
19-131-602 Small
19-131-603 Medium
19-131-604 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-131-605 Supplier PIP™ Supplier No. 348264/XL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves release thousands of micro-encapsulated ingredients, including aloe vera and vitamin E, for positive effects that last long after work hours are over. MicroFoam Nitrile coating is compatible with light oils for enhanced grip capabilities and abrasion resistance.

  • Knit wrist prevents dirt and debris from entering the glove
  • Patented micro-foam nitrile coating offers 360° breathability
  • 25% thinner than most foam nitrile gloves on the market, while offering twice the mechanical performance
  • Glove fit mimics the hand at rest to reduce hand fatigue and increase comfort
  • Constructed using the latest knitting technology to produce smooth and rounded fingertips for increased sensitivity
  • Micro-cup finish allows for a controlled grip in light oil applications
  • Gloves are laundered prior to final packaging to ensure that they are clean and safe for use
  • All ingredients used in the manufacturing and construction of this product are compliant with REACH legislation
  • Oeko-Tex™ Association has evaluated and certified this product as skin safe from the second it comes into contact with the skin
  • Skin Health Alliance has awarded professional dermatological accreditation following review of the scientific dossier behind this product
  • 100% silicone free

Specifications

ANSI Abrasion Rating ANSI 3
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile MicroFoam
Color Pink
Exterior Finish Fully Textured
Hand Specific
Material Nylon, Lycra
Product Type Seamless Knit Gloves
Size X-Large
Style Standard
For Use With (Application) General Assembly, Micro-Engineering, Automotive Components, Painting, and Horticulture
Glove Cuff Style Continuous Knit
Series 34-8264
Yarn Gauge 15
Length (English) 8.9 in.
Length (Metric) 22.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.