Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Maximum Safety™ Cowskin Leather Palm Anti-Vibration Gloves

Catalog No. 19121224D Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-121-224D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-121-224D Supplier PIP™ Supplier No. 122AV40/XL
This item is not returnable. View return policy

The glove's ergonomic design, with a heavy-duty leather palm/back, reduces hand fatigue while offering dexterity and durability.

  • Palm pad insert absorbs and dampens shock to the joints of the hand
  • Cotton terry cloth back for breathability
  • Wrist support protects hands against injury from repetitive movements
  • Half-finger design greater dexterity, sensitivity, and breathability
  • Wrap around wrist support with hook & loop closure for secure fit, easy on/off
  • Launderable fabric extends glove life and reduces replacement costs

Specifications

Color Black
Hand Specific
Material Cowhide Leather
Product Type Anti-vibration Gloves
Size X-Large
Finger Style Half
For Use With (Application) Lifting, Material Handling, and Electric Tool Use
Glove Cuff Style Hook and Loop
Series 122-AV40
Length (English) 8.7 in.
Length (Metric) 22 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.