Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Heavy Weight Seamless Knit Cotton/Polyester Gloves with Double-Sided PVC Dot Grip

Catalog No. 19046229 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-046-229 Small
19-046-129 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-046-229 Supplier PIP™ Supplier No. 36C330PDD/S
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Seamless knit shell is comfortable and flexible, while the PVC dots & coated fingertips enhance grip capabilities and abrasion resistance.

  • Double-sided PVC dots provide enhance grip, while also providing cut and abrasion resistance
  • Coated finger tips extend glove life, and increase durability in high-wear areas
  • Seamless knit shell is comfortable and flexible
  • Knit wrist helps prevent dirt and debris from entering the glove

Specifications

Construction Seamless Knit
Coating Material PVC Dots (2 Sides)
Color Natural
Exterior Finish No Texture
Hand Ambidextrous
Material Polycotton
Product Type Seamless Knit Gloves
Size Small
Style Standard
For Use With (Application) Warehousing, General Assembly, Construction, and Bottle Handling
Glove Cuff Style Continuous Knit
Length (English) 8.9 in.
Length (Metric) 22.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.