Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ PermFlex™ Rough PVC-Coated Cold Resistant Gloves - 10 in.

Catalog No. 19023120B Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-023-120B Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-023-120B Supplier PIP™ Supplier No. 588650/M

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Textured PVC gloves remain soft and flexible in cold applications, while also enhancing grip capabilites. Seamless cotton liner provides tactile sensitivity and is treated to resist germs/odors.

  • PVC material remains soft and flexible in cold applications
  • 10 in. gauntlet cuff provides additional forearm protection
  • Rough-textured palm enhances grip capabilities
  • Seamless cotton liner increases tactile sensitivity
  • Liner is treated to resist germs and odors
  • Ideal for applications that require a combination of liquid and cut protection

Specifications

Coating Material PVC
Color Orange
Exterior Finish Fully Textured
Hand Specific
Material PVC
Product Type Cold-resistant Gloves
Size Medium
For Use With (Application) Cold Water, Marine Biology, and Oily Material Handling
Glove Cuff Style Gauntlet
Liner Material Cotton
Series 58-8650
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.