Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ProCoat™ Premium PVC Coated Yellow Gloves with Interlock Liner - 12 in.

Catalog No. 19823836 Shop All Protective Industrial Products Inc. Products
Click to view available options
:
Large
This item is not returnable. View return policy

Catalog No. 19-823-836

Supplier: PIP™ 588030Y

This item is not returnable. View return policy

Single dip construction with a soft jersey liner for comfort and moderate insulation. Straight cuff adds additional length to protect the forearm from potentially hazardous liquid run-off.

  • Yellow color improves visibility and allows glove degradation to be easily identified
  • Soft jersey liners provide comfort and moderate insulation
  • Straight cuff adds additional length to protect the forearm from liquid run-off

Specifications

Coated Area Full Glove
Coating Material PVC
Color Yellow
Exterior Finish No Texture
Hand Specific
Product Type Dip-coated Gloves
Size Large
For Use With (Application) Mining, Construction, Refining, Petrochemicals, and Oily Material Handling
Glove Cuff Style Gauntlet
Liner Material Cotton
Series 58-8030Y
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.