Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Seamless Knit Cotton/Polyester Gloves with Black PVC Coating

Catalog No. 19121229A Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-121-229A Small
19-121-228A Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-121-229A Supplier PIP™ Supplier No. 37C110PCBK/S
  • $644.00
    $320.65 / Case of 240

    Save $323.35

    50% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Seamless knit construction is comfortable and flexible, while PVC coating on the palm enhances grip capabilities.

  • Seamless knit shell is comfortable and flexible
  • Knit wrist helps prevent dirt and debris from entering the glove
  • PVC coating on the palm enhances grip and provides resistance to cuts and abrasion

Specifications

Chemotherapeutic Rated No
Coated Area Palm
Construction Seamless Knit
Coating Material PVC
Color Black, Natural
Exterior Finish Grip Pattern
Hand Specific
Material Polycotton
Product Type Seamless Knit Gloves
Size Small
For Use With (Application) Package Handling, Warehousing, General Assembly, and Construction
Gender Unisex
Glove Cuff Style Continuous Knit
Liner Material Nylon
Series 37-C110PC
Yarn Gauge 7
Length (English) 8.9 in.
Length (Metric) 22.5 cm
Show More Show Less

WARNING: Cancer - www.P65Warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.