Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Regular Grade Split Cowhide Leather Drivers Gloves

Catalog No. 19167832D Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-167-832D X-Large
19-167-832A Small
19-167-832B Medium
19-167-832C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-167-832D Supplier PIP™ Supplier No. 69134/XL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Split leather is economical and has visible texture. Cowhide provides comfort, durability, abrasion resistance, and breathability.

  • Drivers gloves feature slip-on styling to provide good fit and comfort
  • Elastic closure sewn inside glove for safety and comfort
  • Straight thumb increases comfort in closed-fisted work
  • Color coded hems for easy size identification
  • For use in construction, machine operation, utility work, farming, roping, and many other general applications

Specifications

Color Tan
Grade Regular
Hand Specific
Material Shoulder Split Cowhide Leather
Product Type Driver Gloves
Size X-Large
Style Standard
Glove Cuff Style Open
Hem Color Blue
Series 69-134
Thumb Style Straight
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.