Learn More
PIP™ The Scout™ Rimless Safety Glasses with Clear Temple and Clear Lens
Catalog No. 19161906 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Packaging:
12-Pair Dispenser Box
No Dispenser Box
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
19-161-906 12-Pair Dispenser Box
19-161-644 No Dispenser Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-161-906 Supplier PIP™ Supplier No. 250990980DP
  • $84.00
    $53.65 / Pack of 12
    Save $30.35
    36% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $868.00
    $559.65 / Case of 12 PK
    Save $308.35
    36% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Our most popular visitor's glasses feature a large frame that can be used alone, or as an Over-The-Glass (OTG) style to fit all personal eyewear.
  • Polycarbonate unilens
  • Over-the-glass design fits over all prescription eyewear
  • Can also be worn by itself as a regular safety glass
  • Molded nose bridge design to fit the vast majority of users
  • Indirect vent temples to reduce fogging
  • Clear Lens - clear, crisp and distortion free visibility while meeting the requirement standards that help provide protection from projectiles and impact in general use applications
  • Uncoated
  • Economical style intended to be used by visitors to a facility, or other short-term applications
  • Individually bagged; 12-pair dispenser box
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
Lens Coating Uncoated
Lens Tint Clear
Product Type Safety Glasses
No. per Pack 12/Pk.
Temple Style Vented
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
Frame Type Rimless
Packaging 12-Pair Dispenser Box
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.