Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Stat-Tek™ Women's Nylon ESD Gloves with Urethane Coated Palm

Catalog No. 19121213C Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-121-213C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-121-213C Supplier PIP™ Supplier No. 996457L/L

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lint-free, anti-static nylon with an ultra thin urethane moisture barrier on the palm and thumb.

  • Constructed of Kanebo Belltron conductive fiber and water affinitive Kanebo SLN nylon
  • Absorbs perspiration 50 times better than polyester
  • Lint-free, anti static nylon fabric
  • Ultra thin urethane moisture barrier on the palm and thumb
  • Low particulate count
  • Enhanced comfort and dexterity
  • Ladie's size for true fit on female hands

Specifications

Coated Area Palm
Coating Material Polyurethane
Color White
Material Nylon
Product Type ESD Gloves
Size Large
For Use With (Application) Computer Manufacturing, Product Assembly/Finishing/Handling, and Photography Labs
Gender Women's
Glove Cuff Style Plain
Hem Color Pink
Series 99-6457L
Length (English) 8.5 in.
Length (Metric) 21.6 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.