Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Medium Weight Seamless Knit Cotton/Polyester Gloves

Catalog No. 191201498 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-120-1498 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-120-1498 Supplier PIP™ Supplier No. 35CB110/L
  • $29.10
    $17.65 / Dozen of 12

    Save $11.45

    39% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $727.60
    $441.65 / Case of 25 DZ

    Save $285.95

    39% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Seamless knit cotton/polyester, 7 gauge shell is comfortable and flexible. The knit wrist helps prevent dirt and debris from entering the glove.

  • Reversible design allows replacing one glove at a time, rather than pairs
  • Can also be used as a glove liner
  • Small size is an ideal fit for women's use
  • Made from component materials which comply with federal regulations for food contact, 21 CFR, 170-199

Specifications

Certifications/Compliance 21 CFR, 170-199
Construction String Knit
Description Medium Weight
Accelerator Free Yes
Applications Industrial
Coating Material Uncoated
Color White
Disposable Reusable
Exterior Finish No Texture
Hand Ambidextrous
Material Polycotton
Powdered Powder Free
Product Type General Purpose Gloves
Size Large
Sterility Non-sterile
Style Reversible
Glove Cuff Color White
Glove Cuff Style Continuous Knit
Hem Color White
Yarn Gauge 7
Length (English) 10 in.
Length (Metric) 25 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.