Learn More
Echelon Biosciences Research Labs PIP STRIPS - 2 pack
SDP
Supplier:  Echelon Biosciences Research Labs P60012PK
Encompass
PIP Strips are 2 x 6 cm hydrophobic membranes that have been spotted with 100 pmol of all eight phosphoinositides and seven other biological important lipids. They are used to determine how your protein of interest interacts with one or more of these lipids in a simple protein-lipid overlay assay. Bound protein is then detected by an antibody against the protein or affinity tag. This item is non returnable
Catalog No. 50-143-990
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.