Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ProCoat™ Hi-Vis PVC Coated Insulated Gloves

Catalog No. 19816867 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-816-867 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-816-867 Supplier PIP™ Supplier No. 587303
Add to Cart
Add to Cart
This item is not returnable. View return policy

Foam insulation keeps hands warm and dry, while the highly visible orange color increases visibility in low light applications.

  • PVC coating provides resistant to liquids, as well as increasing grip capabilities
  • Hi-vis orange color improves visibility in warehouses and on highways
  • Triple layered with interlock, foam insulation, and a jersey liner
  • Foam insulation helps to keep hands warm and dry
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Ideal for handling grease, oils, acids, caustics, and solvents in cold environments

Specifications

Coating Material PVC
Color Hi-vis Orange
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material PVC
Product Type Dip-coated Gloves
Size Large
For Use With (Application) Chemical Handling, Construction, Traffic Control, and Warehousing
Glove Cuff Color Black
Glove Cuff Style Knit
Liner Material Foam Insulation
Series 58-7303
Length (English) 11.2 in.
Length (Metric) 28.5 cm
Show More Show Less

WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.