Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Seamless Knit Thermax™ Gloves - 13 Gauge
GSA_VA

Catalog No. 19013600 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-013-600 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-013-600 Supplier PIP™ Supplier No. 41001WL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Thermax™ helps keep moisture away from the hands to keep them warm, dry, and comfortable. Can be worn alone, or used as glove liners in cold weather applications.

  • Thermax™ keeps moisture away to help hands remain warm and dry
  • Can be worn alone, or used as a glove liner in cold weather applications
  • Seamless knit construction ensures a comfortable fit
  • Ambidextrous pattern permits glove to be worn on either hand for extended glove life and reduced replacement costs
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Quickly evaporates moisture from the skin
  • Extra softness for non-chafing comfort
  • Machine washable

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Construction Seamless Knit
Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Latex Free Yes
Material Polyester
Product Type Inspection Gloves
Size Large
Sterility Non-sterile
For Use With (Application) Construction, Commercial Fishing, Food Processing, Refrigeration, and Utility Work
Glove Cuff Style Knit
Liner Material Thermax™
Series 41-001
Yarn Gauge 13
Length (English) 6.9 in.
Length (Metric) 17.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.