Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Base35™ Three-Piece Rainsuit

Catalog No. 191305498 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
5X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-5498 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-130-5498 Supplier PIP™ Supplier No. 201370X5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for construction, municipalities, shipyards, agriculture chemical spraying and industrial plant maintenance.

  • Detachable hood with drawstring
  • Storm flap front, two flapped pockets and underarm vents
  • Adjustable snaps at wrists and ankles for better fit
  • Made with .35mm thick PVC and polyester fabric
  • Bib overalls for full moisture protection
  • Self material collar for easy cleaning
  • Cape vented back
  • Not Compliant with California Proposition 65

Specifications

Color Yellow
For Use With (Application) Construction, municipalities, shipyards, agriculture chemical spraying and industrial plant maintenance
Hood Type Detached
Material Polyester, PVC
Size 5X-Large
No. of Pockets 2
Product Type Three-Piece Rainsuit
Size (English) Chest 66 in.
Size (English) Waist 58 in.
Size (Metric) Chest 167.6cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.