Learn More
PIP™ XtraTuff™ Oil Resistant 10 in. PVC Gloves with Seamless Liner and Rough Coating
Great for applications where machine oil is present and for applications that require a combination of liquid and cut protection.
Manufacturer:  PIP™ 588656/S
 View more versions of this product
Catalog No. 19-175-108
Add to cart
Description
Description
  • Oil resistant, staying soft and strong even after exposure to machine oils
  • Gauntlet cuff for added forearm protection and allows for easy on and off
  • Rough coated palm provides a great grip
  • Seamless liner provides excellent tactile sensitivity and is treated to resist germs and odors
  • Compliant with FDA Part 177 - Indirect food activities: Polymers. Subpart C - Substances for use only as components of articles intended for repeated use. Sec. 177.2600
Specifications
Specifications
Rough
Treated to resist germs/odors
PVC (Polyvinyl Chloride)
Blue
30.5cm
Gauntlet
Interlock knit
Small
12 in.