Learn More
PIP™ Zenon Z-Lyte II™ Rimless Safety Glasses
Catalog No. 19824205 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
Indoor/Outdoor
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
Frame Color:
Clear
Gray
Shape:
Wraparound Lens
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Lens Coating Frame Color Shape
19-824-205 Gray Anti-fog, Scratch-resistant Clear -
19-824-201 Clear Scratch-resistant Clear -
19-824-202 Gray Scratch-resistant Gray -
19-824-204 Clear Anti-fog, Scratch-resistant Clear -
19-824-203 Indoor/Outdoor Scratch-resistant Clear -
19-148-032 Clear Anti-fog, Scratch-resistant Clear Wraparound Lens
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Catalog No. 19-824-205 Supplier PIP™ Supplier No. 250090021
Add to Cart
Add to Cart
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Provides unparalleled comfort and styling. Rimless Safety Glasses features thin temple that provides secure fit without pressure and modern wraparound lens style.
  • Lightweight safety eyewear
  • Wraparound lens style
  • Thin temple provides secure fit without pressure
  • Variety of lens options for virtually any application
  • Squared lens shape with slight taper, provides modern styling
  • Enhanced lens design extends field of vision and protection
  • Sleek temples with strategically perforated design ensures flexible, secure fit to multiple head shapes and sizes
Compliance
Meets ANSI Z87.1-2015 standards.
Specifications
Lens Coating Anti-fog, Scratch-resistant
Description Squared lens with slight taper
Lens Tint Gray
Product Type Safety Glasses
For Use With (Application) Protection from projectiles and impact in general use
Frame Color Clear
Temple Style Thin
Weight (Metric) 16 g
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection 99.99% UVA and UVB Light Absorption
Frame Type Rimless
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.