Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIP5K1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115760
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115760 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115760 Supplier Invitrogen™ Supplier No. PA5115760
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PIP5K1C.

PIP5K1C lipid kinase or phosphatidylinositol-4-phosphate 5-kinase 1 gamma phosphorylates phosphatidylinositol-4-phosphate to form phosphatidylinositol 4,5-bisphosphate. PIP5K1C is found at synapses and has been found to play roles in endocytosis and cell migration. PIP5K1C directly associates with -beta-catenin and increases -beta-catenin activity downstream of growth factor stimulation. PIP5K1C expression and kinase activity enhance -beta-catenin phosphorylation on residues that promote nuclear importation and transcriptional activity. PIP5K1C deficiency impairs neutrophil recruitment as a result of defect in adhesion. PIP5K1C regulated the adhesion through facilitation of RhoA GTPase and integrin activation by chemo attractants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIP5K1C
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene PIP5K1C
Gene Accession No. O60331
Gene Alias AI115456; AI835305; diphosphoinositide kinase; Kiaa0589; LCCS3; phosphatidylinositol 4-phosphate 5-kinase type I gamma; Phosphatidylinositol 4-phosphate 5-kinase type-1 gamma; phosphatidylinositol-4-phosphate 5-kinase; phosphatidylinositol-4-phosphate 5-kinase type 1 gamma; phosphatidylinositol-4-phosphate 5-kinase type I gamma; phosphatidylinositol-4-phosphate 5-kinase type-1 gamma; phosphatidylinositol-4-phosphate 5-kinase, type 1 gamma; phosphatidylinositol-4-phosphate 5-kinase, type I, gamma; Pip5k1c; PIP5K1-gamma; PIP5Kgamma; PIP5K-GAMMA; PIP5KI gamma; Pip5kIgamma; PIP5KI-gamma; PIPKIg_v4; ptdIns(4)P-5-kinase 1 gamma; ptdIns(4)P-5-kinase gamma; ptdInsPKIgamma; type I PIP kinase
Gene Symbols PIP5K1C
Host Species Rabbit
Immunogen A synthesized peptide derived from human PIP5K1C(Accession O60331), corresponding to amino acid residues R304-Y354.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23396
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.