Learn More
Vikan™ Pipe Brush with Handle, One Piece, 3.1 in.
Catalog No. 17200299 Shop All Remco Products
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Purple
Red
White
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-200-299 Green
17-200-297 Red
17-200-294 Orange
17-200-295 Yellow
17-200-298 Blue
17-200-293 Purple
17-200-292 Black
17-200-296 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-200-299 Supplier Vikan™ Supplier No. 5382802
Add to Cart
Add to Cart
The one piece construction and high-quality materials make the pipe cleaning brush durable for repetitive tough cleaning.
  • More bristles holds even cleaning solution
  • Stiff bristles allow the brush to handle tougher cleaning jobs
  • Ergonomic designed
  • Long handle that force can be easily applied
  • The lack of crevices, cracks, and mold lines eliminates the places bacteria can hide and multiply in
Specifications
Product Type Pipe Brush
Bristle Material PBT
Bristle Type Medium Stiffness
Color-coded Yes
Handle Material Polypropylene
Handle Type Ergonomic
Tip Type Without Tip
Sterility Non-sterile
Color Green
Length (English) 3.1 in.