Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIG™ Pipe Leak Diverter Tarp
SDP

Catalog No. 18366646 Shop All New Pig Corporation Products
Change view
Click to view available options
Material:
Polyester-blended PVC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
18-366-646 Polyester-blended PVC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-366-646 Supplier PIG™ Supplier No. TLS691YW
Add to Cart
Add to Cart

Pipe leak diverter trap that quickly installs to channel away pipe leaks before it damages inventory and machinery or causes a slip hazard.

  • 24 in. x 24 in. leak diverter tarp
  • Hangs from a leaking pipe to catch leaks and channel them to a drain or collection container
  • Keeps rain and snow from coming inside
  • Heavy-duty vinyl-coated tarp resists punctures, UV and mildew to last multiple deployments
  • Built-in brass grommets and included rubber straps make it easy to hang from an overhead pipe
  • Brass grommets resist tearing; multiple hanging points add stability and preserve funnel shape
  • Flow rate of 3.33 gal./min. when used with a 1/2 in. ID hose
  • Hose and collection container not included

Specifications

Color Yellow
Material Polyester-blended PVC
Type Roof Leak Diverter Tarp
Height (English) 24 in.
Height (Metric) 61 cm
Diameter (English) 24 in.
Diameter (Metric) 61 cm
Weight (English) 2.43 lb.
Weight (Metric) 1.1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.