Learn More
New Pig Pipe Leak Diverter Tarp
SDP
Catalog No. 18389150 Shop All New Pig Corporation Products
Change view
Click to view available options
Material:
PVC
Catalog No. Material
18-389-150 PVC
1 options
This item is not returnable. View return policy
Catalog No. 18-389-150 Supplier New Pig Supplier No. TLS248YW
Add to Cart
Add to Cart
This item is not returnable. View return policy
Catches pipe leaks or condensation to protect equipment, inventory and people.
  • Easily keep rain and snow from storming inside. Whether condensation and ice, or heavy rain from a high-wind hurricane, this solution will quickly protect your team and what you've built
  • Hangs from a leaking pipe to catch leaks and channel them to a drain or collection container
  • Heavy-duty vinyl-coated tarp resists punctures, UV and mildew and lasts multiple deployments
  • Built-in brass grommets and included rubber straps make it easy to hang from an overhead pipe; multiple hanging points add stability and preserve funnel shape
  • Choose yellow for high visibility, gray to blend with plant ceilings or clear to monitor leaks
  • Flow rate of 3.33 gallons per minute when used with a 1/2 in. ID hose
Specifications
Material PVC
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov