Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Pipe Marker for Gas, Legend: BREATHING AIR EPMTY CYLINDERS

Catalog No. 19112052 Shop All Brady Corporation Products
Catalog No. 19-112-052 Supplier Brady™ Supplier No. 17944

For identification of pipe content. Brady chemical and hazardous material signs pipe markers for gases are made from pressure sensitive vinyl with permanent adhesive.

Specifications

Description Abrasion and chemical resistant
Adhesive Type Acrylic, Permanent, Cold-temperature
Color White, Blue
Material Pressure-sensitive Vinyl (B-946)
Product Type Pipe marker for gases
Display Text Legend: BREATHING AIR EPMTY CYLINDERS
Size (English) 10 x 7 in. (W x H)
Size (Metric) 25.4 x 17.7 cm (W x H)
Thickness (English) 0.004 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.