Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pipemidic Acid, G-Biosciences
SCI_ED

Catalog No. S000037
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000037 100 g
S139796 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000037 Supplier G-Biosciences Supplier No. RC1087
Add to Cart
Add to Cart

Pipemidic acid is a pyridopyrimidine antibiotic with similar properties to quinolone antibiotics.

It is used to study damage and mutations in DNA as well as lymphocyte blastogenesis and interferon-gamma production. It can be used as a cell selection agent.

Chemical Identifiers

Molecular Weight (g/mol) 303.32 g/mol

Specifications

Boiling Point 534.9°C
Quantity 100 g
Percent Purity 98%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.