Learn More
Apexbio Technology LLC Piperacillin Sodium 59703-84-3 10mM (in 1mL DMSO)
Supplier: Apexbio Technology LLC B18185.1
Piperacillin Sodium (CAS 59703-84-3) is a small-molecule inhibitor targeting bacterial penicillin-binding proteins (PBPs) It is designed to inhibit cell wall biosynthesis thereby disrupting peptidoglycan cross-linking in bacteria Piperacillin Sodium exerts its biological activity primarily through irreversible binding to bacterial transpeptidases (PBPs) In in vitro studies Piperacillin Sodium demonstrates antibacterial inhibition with IC50 values typically ranging from 2 to 16 g/ml depending on microorganism sensitivity and experimental conditions Based on these pharmacological properties Piperacillin Sodium holds research potential in microbiological studies and the investigation of Pseudomonas aeruginosa infections
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.