Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperacillin sodium salt, 50 mg/ml in distilled water, sterile-filtered, Thermo Scientific Chemicals

Catalog No. AAJ67261XF
Change view
Click to view available options
Quantity:
1 mL
5 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67261XF 1 mL
AAJ672618EQ 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67261XF Supplier Thermo Scientific Chemicals Supplier No. J67261XF
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59703-84-3
Molecular Formula C23H26N5NaO7S
Molecular Weight (g/mol) 539.54
MDL Number MFCD00917471
InChI Key WCMIIGXFCMNQDS-QMJSIVKPSA-M
PubChem CID 23666879
ChEBI CHEBI:8233
IUPAC Name sodium (2S,5R,6R)-6-[(2S)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [Na+].CCN1CCN(C(=O)N[C@H](C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C2=CC=CC=C2)C(=O)C1=O

Specifications

Type Piperacillin Sodium Salt
Physical Form Liquid
Packaging Plastic Bottle
Quantity 1 mL
Chemical Name or Material Piperacillin sodium salt
Hazard Category H317-H334
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
EINECSNumber 261-868-6
RTECSNumber XI0180000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.