Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperazine Adipate 98.0+%, TCI America™
SDP

Catalog No. P044825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P044825G 25 g
P0448500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P044825G Supplier TCI America Supplier No. P044825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 142-88-1
Molecular Formula C10H20N2O4
Molecular Weight (g/mol) 232.28
MDL Number MFCD00050650
InChI Key BVEGEKOBSPXUJS-UHFFFAOYSA-N
PubChem CID 8905
IUPAC Name hexanedioic acid;piperazine
SMILES C1CNCCN1.C(CCC(=O)O)CC(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 232.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Piperazine Adipate
RTECSNumber MO1955000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.