Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperazine, Anhydrous, 99%, Spectrum™ Chemical
SDP

Catalog No. 18612383 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-383 2.5 kg Amber Glass Bottle
18-612-385 5 g Amber Glass Bottle
18-612-381 100 g Amber Glass Bottle
18-612-387 500 g Amber Glass Bottle
18-612-382 12 kg Amber Glass Bottle
18-612-384 25 kg Amber Glass Bottle
18-612-386 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-612-383 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PI1232.5KG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 110-85-0
Molecular Formula C4H10N2
Molecular Weight (g/mol) 86.14
InChI Key GLUUGHFHXGJENI-UHFFFAOYSA-N
IUPAC Name piperazine
SMILES C1CNCCN1

Specifications

CAS Max % 1
Melting Point 108° to 113°C
Boiling Point 147°C / 296.6°F
Packaging Amber Glass Bottle
Quantity 2.5 kg
Percent Purity 0.99
Grade Reagent
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Piperazine, Anhydrous

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.