Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperazine hexahydrate, ≥99% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111019092 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
200 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9092 1 kg
11-101-9727 200 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9092 Supplier MilliporeSigma Supelco Supplier No. P70031KG
Add to Cart
Add to Cart

Piperazine hexahydrate may be used as a standard for the determination of piperazine in pharmaceutical preparations using colorimetric technique.

Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 142-63-2
Vapor Pressure 0.8 mmHg (20°C)
Flash Point 88°C
Physical Form Neat
Linear Formula C4H10N2 · 6H2 O
MDL Number MFCD00149389
Melting Point 42°C to 44°C
Boiling Point 145°C to 156°C
Type Analytical Standard
Quantity 1 kg
Formula Weight 194.23
Percent Purity ≥99% (TLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.