Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperazine Sulfate Hydrate, TCI America™
SDP

Catalog No. P045225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P045225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P045225G Supplier TCI America Supplier No. P045225G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4554-26-1
Molecular Formula C4H12N2O4S
Molecular Weight (g/mol) 184.21
MDL Number MFCD00060186
InChI Key MYNIYCGOBKAQAO-UHFFFAOYSA-N
PubChem CID 3014216
IUPAC Name piperazine; sulfuric acid
SMILES OS(O)(=O)=O.C1CNCCN1

Specifications

Color White
Quantity 25 g
Formula Weight 184.21
Physical Form Crystalline Powder
Chemical Name or Material Piperazine Sulfate Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.