Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIPES, 99.4%, MP Biomedicals™

Catalog No. ICN19025790 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19025790 500 g
ICN19025710 10 g
ICN19025780 100 g
ICN19025791 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19025790 Supplier MP Biomedicals, Inc Supplier No. 0219025790
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5625-37-6
Molecular Formula C8H18N2O6S2
Molecular Weight (g/mol) 302.36
InChI Key IHPYMWDTONKSCO-UHFFFAOYSA-N
Synonym Piperazine-N, N'-bis[2-ethanesulfonic acid]
PubChem CID 79723
ChEBI CHEBI:44933
IUPAC Name 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
SMILES C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 0.994
Formula Weight 302.4
Chemical Name or Material Pipes
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.