Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIPES Sesquisodium Salt, MP Biomedicals™

Catalog No. 0215245610 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-152-45610 10 g
ICN15245680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-152-45610 Supplier MP Biomedicals, Inc Supplier No. 0215245610
Add to Cart
Add to Cart

PIPES Sesquisodium Salt is a zwitterionic buffer.

  • PIPES is frequently used as a buffering agent in biochemistry.
  • It is an ethanesulfonic acid buffer developed by Good et al. in the 1960s.
  • PIPES has a pKa near the physiological pH which makes it useful in cell culture work.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100037-69-2
Molecular Formula C16H33N4Na3O12S4
InChI Key FPDACHMSOMILQI-UHFFFAOYSA-K

Specifications

Quantity 10 g
SMILES [Na+].[Na+].[Na+].OS(=O)(=O)CCN1CCN(CCS([O-])(=O)=O)CC1.[O-]S(=O)(=O)CCN1CCN(CCS([O-])(=O)=O)CC1
IUPAC Name trisodium 2-[4-(2-sulfoethyl)piperazin-1-yl]ethane-1-sulfonate 2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethane-1-sulfonate
Molecular Weight (g/mol) 670.67

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.