accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNaOHSOONNSOOOOSOONNSOOO

CAS RN 100037-69-2

IUPAC Name: trisodium 2-[4-(2-sulfoethyl)piperazin-1-yl]ethane-1-sulfonate 2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethane-1-sulfonate
Synonyms: trisodium 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonate 2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethanesulfonate
Molecular Weight (g/mol): 670.67
Molecular Formula: C16H33N4Na3O12S4
InChi Key: FPDACHMSOMILQI-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[Na+].OS(=O)(=O)CCN1CCN(CCS([O-])(=O)=O)CC1.[O-]S(=O)(=O)CCN1CCN(CCS([O-])(=O)=O)CC1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Physical Form 
  • (1)
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)