Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIPES, Sesquisodium Sodium Salt, Fisher BioReagents

Catalog No. BP304500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP304-500 500 g Poly Bottle
BP304-100 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BP304-500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Commonly used buffering agent

This biological buffer has a usable pH range of 6.1 to 7.5.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100037-69-2
Molecular Formula C16H33N4Na3O12S4
Molecular Weight (g/mol) 670.665
InChI Key FPDACHMSOMILQI-UHFFFAOYSA-K
Synonym 1, 4-Piperazinediethanesulfonic acid sesquisodium salt, Piperazine-1, 4-bis(2-ethanesulfonic acid) sesquisodium salt
PubChem CID 3086462
IUPAC Name trisodium;2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonate;2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethanesulfonate
SMILES C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)[O-].C1CN(CCN1CCS(=O)(=O)[O-])CCS(=O)(=O)[O-].[Na+].[Na+].[Na+]

Specifications

Melting Point 300°C
Color White
pH 6.5 to 7.1
Physical Form Powder or Solid
Quantity 500 g
DNase DNase free
Identification Pass Test
Assay Percent Range ≥95 %
Packaging Poly Bottle
Protease Protease free
Formula Weight 670.68
Percent Purity ≥95%
Grade DNase- and RNase-Free
Chemical Name or Material PIPES, Sesquisodium Salt
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.